CAS RN 100037-69-2
CAS RN 100037-69-2
IUPAC Name:
trisodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms:
trisodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
Molecular Weight (g/mol):
670.67
Molecular Formula:
C16H33N4Na3O12S4
InChi Key:
FPDACHMSOMILQI-UHFFFAOYSA-K
SMILES:
[Na+].[Na+].[Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1.[O-]S(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
1
–
2
of
2
results
1
PIPES Sesquisodium Salt, MP Biomedicals™
CAS: 100037-69-2 Molecular Formula: C16H33N4Na3O12S4 InChI Key: FPDACHMSOMILQI-UHFFFAOYSA-K Synonym: 1, 4-Piperazinediethanesulfonic acid sesquisodium salt,Piperazine-1, 4-bis(2-ethanesulfonic acid) sesquisodium salt PubChem CID: 3086462
2
PIPES, Sesquisodium Sodium Salt, Fisher BioReagents
CAS: 100037-69-2 Molecular Formula: C16H33N4Na3O12S4 Molecular Weight (g/mol): 670.665 InChI Key: FPDACHMSOMILQI-UHFFFAOYSA-K Synonym: 1, 4-Piperazinediethanesulfonic acid sesquisodium salt,Piperazine-1, 4-bis(2-ethanesulfonic acid) sesquisodium salt PubChem CID: 3086462 IUPAC Name: trisodium;2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate SMILES: C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)[O-].C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+].[Na+]