accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10043-67-1

CAS RN 10043-67-1

IUPAC Name: aluminium(3+) potassium disulfate
Synonyms: potassium alum
Molecular Weight (g/mol): 258.19
Molecular Formula: AlKO8S2
InChi Key: GRLPQNLYRHEGIJ-UHFFFAOYSA-J
SMILES: [Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.