accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10049-01-1

CAS RN 10049-01-1

IUPAC Name: bismuth(3+) phosphoric acid
Synonyms: bismuth(3+) phosphoric acid
Molecular Weight (g/mol): 306.97
Molecular Formula: BiH3O4P
InChi Key: SFOQXWSZZPWNCL-UHFFFAOYSA-N
SMILES: [Bi+3].OP(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.