accessibility menu, dialog, popup

Test

CAS RN 10049-07-7

CAS RN 10049-07-7

IUPAC Name: rhodium(3+) trichloride
Synonyms: rhodium(III) chloride
Molecular Weight (g/mol): 209.26
Molecular Formula: Cl3Rh
InChi Key: SONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)