accessibility menu, dialog, popup

Test

CAS RN 1005-39-6

H3CSNNH2H2NN

CAS RN 1005-39-6

IUPAC Name: 2-(methylsulfanyl)pyrimidine-4,6-diamine
Synonyms: 2-(methylsulfanyl)pyrimidine-4,6-diamine
Molecular Weight (g/mol): 156.21
Molecular Formula: C5H8N4S
InChi Key: AHAIUNAIAHSWPG-UHFFFAOYSA-N
SMILES: CSC1=NC(N)=CC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results