accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10101-58-3

CAS RN 10101-58-3

IUPAC Name: λ²-cobalt(2+) tungsten tetraoxidandiide
Synonyms: λ²-cobalt(2+) tungsten tetraoxidandiide
Molecular Weight (g/mol): 306.77
Molecular Formula: CoO4W
InChi Key: BGLJOHZOBUGBBY-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Co++].[W]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.