accessibility menu, dialog, popup

Test

CAS RN 1017779-49-5

OFFFN

CAS RN 1017779-49-5

IUPAC Name: 4-(difluoromethoxy)-2-fluorobenzonitrile
Synonyms: 4-(difluoromethoxy)-2-fluorobenzonitrile
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H4F3NO
InChi Key: OERIHGQAGTVWPN-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC(F)=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results