accessibility menu, dialog, popup

Test

CAS RN 1017779-88-2

CH3OFOHOF

CAS RN 1017779-88-2

IUPAC Name: 3-ethoxy-2,4-difluorobenzoic acid
Synonyms: 3-ethoxy-2,4-difluorobenzoic acid
Molecular Weight (g/mol): 202.16
Molecular Formula: C9H8F2O3
InChi Key: KUOUFQUWQYELBO-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=CC(C(O)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results