accessibility menu, dialog, popup

Test

CAS RN 1019-57-4

H3CNNHNOONOO

CAS RN 1019-57-4

IUPAC Name: (2E)-1-(2,4-dinitrophenyl)-2-ethylidenehydrazine
Synonyms: (2E)-1-(2,4-dinitrophenyl)-2-ethylidenehydrazine
Molecular Weight (g/mol): 224.18
Molecular Formula: C8H8N4O4
InChi Key: ONBOQRNOMHHDFB-XNWCZRBMSA-N
SMILES: C\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results