man

Test

CAS RN 102-08-9

SHNHN

CAS RN 102-08-9

IUPAC Name: 1,3-diphenylthiourea
Synonyms: thiocarbanilide
Molecular Weight (g/mol): 228.31
Molecular Formula: C13H12N2S
InChi Key: FCSHMCFRCYZTRQ-UHFFFAOYSA-N
SMILES: S=C(NC1=CC=CC=C1)NC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results