accessibility menu, dialog, popup

UserName

CAS RN 102-71-6

OHNHOOH

CAS RN 102-71-6

IUPAC Name: 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: triethanolamine triethanolamine citrate (1:1) caswell no. 886 daltogen triethanolamine maleate sting-kill nitrilo-2,2',2''-triethanol triethanolamine iodohydrate nitrilotriethanol triethanolamine sulfite (1:1)
Molecular Weight (g/mol): 149.19
Molecular Formula: C6H15NO3
InChi Key: GSEJCLTVZPLZKY-UHFFFAOYSA-N
SMILES: OCCN(CCO)CCO
Molecular Weight (g/mol) 
  • (8)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (8)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)