accessibility menu, dialog, popup

Test

CAS RN 10205-34-2

H3COOCH3NHCH3CH3

CAS RN 10205-34-2

IUPAC Name: [(2R)-3-methoxy-2-methyl-3-oxopropyl]dimethylazanium
Synonyms: [(2R)-3-methoxy-2-methyl-3-oxopropyl]dimethylazanium
Molecular Weight (g/mol): 146.21
Molecular Formula: C7H16NO2
InChi Key: DDLJXUCYCFDNCA-ZCFIWIBFSA-O
SMILES: COC(=O)[C@H](C)C[NH+](C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results