accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now
Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 122536-76-9

CAS RN 122536-76-9

IUPAC Name: tert-butyl N-(pyrrolidin-3-yl)carbamate
Synonyms: tert-butyl N-(pyrrolidin-3-yl)carbamate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: DQQJBEAXSOOCPG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCNC1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.