CAS RN 256925-03-8
CAS RN 256925-03-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AL 082D06, MedChemExpress
MedChemExpress AL 082D06 is a selective, nonsteroidal glucocorticoid receptor (GR) antagonist with Ki of 210 nM.