accessibility menu, dialog, popup

UserName

CAS RN 304858-67-1

BFNHOOH

CAS RN 304858-67-1

IUPAC Name: (3-cyano-5-fluorophenyl)boronic acid
Synonyms: 3-cyano-5-fluorophenylboronic acid
Molecular Weight (g/mol): 164.93
Molecular Formula: C7H5BFNO2
InChi Key: DLYWCECHXBOCAS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(F)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results