CAS RN 308362-25-6
CAS RN 308362-25-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ridinilazole, MedChemExpress
MedChemExpress Ridinilazole is a novel antibacterial with MICs range of 0.06-0.25 μg/mL (MIC90=8 μg/mL) against C. difficile.