CAS RN 3621-36-1
CAS RN 3621-36-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Columbamine, MedChemExpress
MedChemExpress Columbamine is a quaternary isoquinoline alkaloid isolated from Argemone mexicana.