CAS RN 482333-74-4
CAS RN 482333-74-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Saccharin 1-methylimidazole, MedChemExpress
MedChemExpress Saccharin 1-methylimidazole is an activator for DNA/RNA Synthesis.