CAS RN 58473-74-8
CAS RN 58473-74-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cinromide, MedChemExpress
MedChemExpress Cinromide is an anticonvulsant agent. Cinromide inhibits epithelial neutral amino acid transporter B0AT1 (SLC6A19) with an IC50 of 0.5 μM.