accessibility menu, dialog, popup

UserName

CAS RN 589-08-2

H3CNH

CAS RN 589-08-2

IUPAC Name: methyl(2-phenylethyl)amine
Synonyms: benzeneethanamine, N-methyl-
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: SASNBVQSOZSTPD-UHFFFAOYSA-N
SMILES: CNCCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results