accessibility menu, dialog, popup

UserName

CAS RN 6482-24-2

CH3OBr

CAS RN 6482-24-2

IUPAC Name: 1-bromo-2-methoxyethane
Synonyms: 2-bromoethyl methyl ether
Molecular Weight (g/mol): 138.99
Molecular Formula: C3H7BrO
InChi Key: YZUPZGFPHUVJKC-UHFFFAOYSA-N
SMILES: COCCBr
Molecular Weight (g/mol) 
  • (7)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (4)
  • (4)
Percent Purity 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (4)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)