accessibility menu, dialog, popup

UserName

CAS RN 75-26-3

CH3CH3Br

CAS RN 75-26-3

IUPAC Name: 2-bromopropane
Synonyms: 2-bromopropane
Molecular Weight (g/mol): 122.99
Molecular Formula: C3H7Br
InChi Key: NAMYKGVDVNBCFQ-UHFFFAOYSA-N
SMILES: CC(C)Br
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (6)