accessibility menu, dialog, popup

UserName

CAS RN 924296-39-9

ClONNNN

CAS RN 924296-39-9

IUPAC Name: 7-chloro-9-oxo-9H-indeno[1,2-b]pyrazine-2,3-dicarbonitrile
Synonyms: 7-chloro-9-oxoindeno[1,2-b]pyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 266.64
Molecular Formula: C13H3ClN4O
InChi Key: BIGPXXAUSQLTQR-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(C=C1)C1=NC(C#N)=C(N=C1C2=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results