man

Test

CAS RN 97426-53-4

CH3SH2N

CAS RN 97426-53-4

IUPAC Name: 2-(4-methylphenyl)ethanethioamide
Synonyms: 2-(4-methylphenyl)ethanethioamide
Molecular Weight (g/mol): 165.25
Molecular Formula: C9H11NS
InChi Key: NDIXABMZJKQGHJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC(N)=S)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results