missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Spare Glass Bottle

Buerkle™ Spare Glass Bottle for use with Buerkle™ Dipping Bottle.

Brand:  Buerkle™ 5365-4020

32.17 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


Product Code. 11836403

  • 38.55 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With the Easy-Click
  • Safe and practical closure
  • Easy and rapid to exchange the bottle
  • Spare 1000mL bottle
  • For use with dripping bottle
Specifications

Specifications

Replacement article for Dipping Bottle
1000 mL
110 mm
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.