missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ V2A Stainless Steel Scoop
Buerkle™ V2A Stainless Steel Scoop has no openings, undercuts or crevices in which residues could collect.
Brand: Buerkle™ 5354-1000
This item is not returnable.
View return policy
Description
- Scoop made of stainless steel AISI 304 (1.4301), complete with rod
- Sterilisable by flame sterilisation or steam sterilisation
- Pore-free finishing produced by high-quality laser welding seams
Specifications
Scoop | |
Autoclavable | |
1000 mL |
Stainless Steel (V2A) | |
Sterilisable by flame or steam sterilisation | |
115 cm |