Learn More
0.1 NTU Ratio Turbidity Standard, Reagecon
BIOPHARMA
A suspension of polymer microspheres designed to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards
Brand:  Reagecon CRSR-0.1-500
Product Code. 11429159
  • 182.00 € / 500mL
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Guaranteed stability throughout shelf life even after opening bottle
  • ± 1% accuracy
  • NIST traceable
  • US EPA approved
Specifications
Specifications
Ratio Instruments
± 1% NTU
HDPE Bottle GL45
Liquid
500 mL