missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chondrolectin Antibody (396305), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2576P
This item is not returnable.
View return policy
Description
Chondrolectin Monoclonal antibody specifically detects Chondrolectin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Chondrolectin | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 396305 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| C21orf68, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
| Mouse myeloma cell line NS0-derived recombinant human Chondrolectin, Arg22-Asn216, Accession # Q9H9P2 | |
| 0.1 mL | |
| Developmental Biology | |
| 140578 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction