Learn More
Motic™ 0.35X C-Mount Camera Adapter
This C-mount camera adapter allows Moticams to connect with Motic™ trinocular microscopes.
Brand:  Motic™ 1101001904111
Product Code. 15429254
  • 207.00 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Compatible with the following cameras:
  • Moticam 2
  • Moticam X
  • Moticam X3
  • Moticam 1080
  • Moticam BTU 10
  • Moticam BTX 8
Specifications
Specifications
Camera Adapter
Motic™ BA, AE2000/AE31E, SMZ-161/171 Trinocular Microscopes