Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Features chemical resistant O-ring
Brand:  Axygen™ SCT-050-C-S
 View more versions of this product
Product Code. 12709498
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Cap
Clear
0.5 mL
Polypropylene
O-ring
500 Tubes and caps/Pk.; 8 Packs/Cs.
No
Conical
RNase, DNase, pyrogen-free
Polypropylene
Clear
Non-looped
500/Pk.
Sterile
Screw Cap
20,000 x g