missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0129 Discs

Product Code. p-8436008
Click to view available options
Concentration:
150 μg
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10076142

Brand: Thermo Scientific™ DD0015T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Differentiate Vibrios from other Gram-negative rods using these diagnostic discs.

Differentiate vibrios from other Gram-negative rods using Thermo Scientific™ Oxoid™ 0129 Discs. The test exploits the sensitivity of vibrios to the vibrio static agent 0129 (2,4-Diamino-6,7-di-iso-propylpteridine phosphate)1. 0129 Discs are of great value in differentiating vibrios from other Gram-negative rods and particularly from aeromonads, which are characteristically resistant to 0129. The degree of sensitivity of vibrios to 0129 can also be used as a diagnostic feature in differentiation of Vibrio species.

50 Discs/Cartridge.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Concentration 150 μg
Detectable Analytes Vibrio species, Gram-negative rods
Format Disk, Cartridge
Type O129 Disc
Packaging Type 50 Discs/Cartridge
Quantity 150μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.