missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, 99+%

Product Code. 11450713
Click to view available options
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
Questo articolo non è restituibile. Consulta la politica di reso
Normativo:Questo prodotto contiene una sostanza elencata nell'allegato I del regolamento UE n. 517/2014 sui gas fluorurati ad effetto serra. Tutti i clienti devono compilare una dichiarazione prima della consegna.

Codice prodotto. 11450713

missing translation for 'mfr': Thermo Scientific Alfa Aesar A12747.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso
Normativo:Questo prodotto contiene una sostanza elencata nell'allegato I del regolamento UE n. 517/2014 sui gas fluorurati ad effetto serra. Tutti i clienti devono compilare una dichiarazione prima della consegna.

CAS: 920-66-1 | C3H2F6O | 168.038 g/mol

1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

Solubility
Miscible with water, chloroform, ether and acetone.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases, aluminum, zinc and alkali metals.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
MDL Number MFCD00011651
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifica

Melting Point -4°C
Density 1.619
Boiling Point 59°C to 60°C
Linear Formula CF3CH(OH)CF3
Refractive Index 1.275
Quantity 100 g
UN Number UN1760
Beilstein 1841007
Solubility Information Miscible with water,chloroform,ether and acetone.
Formula Weight 168.04
Percent Purity ≥99%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 1,1,1,3,3,3,-Hexafluoro-2-propanol
Avvertenza
  • Danger
Categoria di pericolo
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
Indicazioni di pericolo
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H314-Causes severe skin burns and eye damage.
Consigli di prudenza
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato