missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrachloroethane-d{2}, 99.5% (Isotopic), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
50 g
Description
1,1,2,2-Tetrachloroethane-d_2 is used as an intermediate in chemical research and in pharmaceutics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Liquid |
Color | Colorless |
Density | 1.620 g/mL |
Refractive Index | 1.493 |
Quantity | 2 g |
Melting Point | -44°C |
Boiling Point | 147°C |
Linear Formula | Cl2CDCDCl2 |
UN Number | UN1702 |
Solubility Information | Slightly soluble in water. Soluble in ether,chloroform,ethanol,tetrachloromethane,methanol,acetone,petroleum spirit,carbon disulfide,dimethyl formamide and benzene. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'