missing translation for 'onlineSavingsMsg'
Learn More

1,1,3-Triphenylpropargyl alcohol, 97%, Thermo Scientific Chemicals

Product Code. 15480298
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1522-13-0
Molecular Formula C21H16O
Molecular Weight (g/mol) 284.36
MDL Number MFCD00004449
InChI Key VWRQCJRTHKUVNF-UHFFFAOYSA-N
Synonym 1,1,3-triphenylpropargyl alcohol, 1,1,3-triphenyl-2-propyn-1-ol, diphenyl phenylethynyl carbinol, benzenemethanol, .alpha.-phenyl-.alpha.-phenylethynyl, triphenylprop-2-yn-1-ol, 1,1,3-triphenylpropargylalcohol, 2-propyn-1-ol, 1,1,3-triphenyl, pubchem14025, acmc-209d7i, triphenyl-1-propyne-3-ol
PubChem CID 137058
IUPAC Name 1,1,3-triphenylprop-2-yn-1-ol
SMILES OC(C#CC1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 80°C to 82°C
Quantity 25 g
Formula Weight 284.36
Percent Purity 97%
Chemical Name or Material 1,1,3-Triphenylpropargyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'