missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%, Thermo Scientific Chemicals

Product Code. 15412476
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3083-10-1
MDL Number MFCD00025684

Specifications

Density 0.847
Boiling Point 130°C (11 mmHg)
Flash Point 102°C (216°F)
Molecular Formula C12H30N4
Refractive Index 1.456
Quantity 25 g
UN Number UN3725
Synonym 1,1,4,7,10,10-hexamethyltriethylenetetramine, hexamethyltriethylenetetramine, 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane, hmteta, 1,2-ethanediamine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethyl, n1,n1'-ethane-1,2-diyl bis n1,n2,n2-trimethylethane-1,2-diamine, n,n,n',n,n',n'-hexamethyltriethylenetetramine, triethylenetetramine, 1,1,4,7,10,10-hexamethyl, 1,1,4,7,10,10-hexamethyl-triethylenetetramine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethylethylenediamine
Solubility Information Soluble in water.
InChI Key DWFKOMDBEKIATP-UHFFFAOYSA-N
SMILES CN(C)CCN(C)CCN(C)CCN(C)C
IUPAC Name N'-[2-[2-(dimethylamino)ethyl-methylamino]ethyl]-N,N,N'-trimethylethane-1,2-diamine
Molecular Weight (g/mol) 230.40
PubChem CID 76509
Formula Weight 230.39
Percent Purity 97%
Chemical Name or Material 1,1,4,7,10,10-Hexamethyltriethylenetetramine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.