Learn More
1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%, Thermo Scientific Chemicals
37.59 € - 266.33 €
Chemical Identifiers
CAS | 3030-47-5 |
---|---|
Molecular Formula | C9H23N3 |
Molecular Weight (g/mol) | 173.3 |
MDL Number | MFCD00014876 |
InChI Key | UKODFQOELJFMII-UHFFFAOYSA-N |
Synonym | pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine |
PubChem CID | 18196 |
ChEBI | CHEBI:39475 |
IUPAC Name | N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine |
SMILES | CN(C)CCN(C)CCN(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10453171
|
Thermo Scientific Acros
416950250 |
25 g | Glass bottle |
37.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10279940
|
Thermo Scientific Acros
416951000 |
100 g | Glass bottle |
87.03 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115703
|
Thermo Scientific Acros
416955000 |
500 g | Glass bottle |
266.33 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3030-47-5 | |
173.3 | |
UKODFQOELJFMII-UHFFFAOYSA-N | |
18196 | |
N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine |
C9H23N3 | |
MFCD00014876 | |
pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine | |
CHEBI:39475 | |
CN(C)CCN(C)CCN(C)C |
Specifications
3030-47-5 | |
100.0 | |
White | |
200°C | |
Authentic | |
C9H23N3 | |
1.4415 to 1.4435 | |
0.829 | |
Solubility in water: soluble | |
CN(C)CCN(C)CCN(C)C | |
173.3 | |
CHEBI:39475 | |
173.3 | |
Powder |
99.0 | |
18°C | |
0.8290g/mL | |
72°C | |
98% min. (GC) | |
(CH3)2NCH2CH2N(CH3)CH2CH2N(CH3)2 | |
MFCD00014876 | |
pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine | |
UKODFQOELJFMII-UHFFFAOYSA-N | |
N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine | |
18196 | |
1.52 mPa.s (20°C) | |
98+% | |
1,1,4,7,7-Pentamethyldiethylenetriamine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 221-201-1
- 1,1,4,7,7-Pentamethyldiethylenetriamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only