missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Binaphthyl, 97%, Thermo Scientific Chemicals

Product Code. 10259450
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 604-53-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 143°C to 146°C
Color Brown
Boiling Point >360°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C20H14
MDL Number MFCD00041740
Quantity 5 g
Synonym 1,1'-binaphthalene, 1,1'-binaphthyl, 1,1'-dinaphthyl, binaphthyl, binaphthalene, 1-naphthalen-1-yl naphthalene, 1,1'-binaphtyl, di-.alpha.-naphthol, 1-1-naphthyl naphthalene, di-alpha-naphthol
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ethanol,ether and acetone
InChI Key ZDZHCHYQNPQSGG-UHFFFAOYSA-N
SMILES C1=CC2=CC=CC(C3=CC=CC4=CC=CC=C34)=C2C=C1
IUPAC Name 1-naphthalen-1-ylnaphthalene
Molecular Weight (g/mol) 254.33
PubChem CID 11789
Formula Weight 254.33
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 1'-Binaphthyl, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'