missing translation for 'onlineSavingsMsg'
Learn More
1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals Product Code.: 10553674

1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals

Product Code. 10553674
500 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10553674

Brand: Thermo Scientific Acros 151815000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-62-1
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD00005286
InChI Key PFKFTWBEEFSNDU-UHFFFAOYSA-N
Synonym 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazoleShow MoreShow Less
PubChem CID 68263
IUPAC Name di(imidazol-1-yl)methanone
SMILES O=C(N1C=CN=C1)N1C=CN=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 118.0°C to 120.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Quantity 500 g
Beilstein 23, V,4, 245
Fieser 01,114; 02,61; 05,97; 06,97; 08,77; 09,96; 12,106; 13,66; 16,64
Merck Index 15, 1820
Solubility Information Solubility in water: decomposes. Other solubilities: moderately soluble in thf, soluble in ethanol
Formula Weight 162.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Carbonyldiimidazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464386 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number151810250,151815000,151811000,151810100
Lot NumberA0461553 Certificate TypeCertificate of Analysis Date08/07/2024 Catalog Number151810250,151815000,151811000,151810100
Lot NumberA0457812 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number151810250,151815000,151811000,151810100
Lot NumberA0453442 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number151810250,151815000,151811000,151810100
Lot NumberA0395051 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number151810250,151815000,151811000,151810100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1``-Carbonyldiimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.