missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Cyclohexen-1-yl)pyrrolidine, 97%, Thermo Scientific Chemicals

Product Code. 11431647
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-99-1
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00003163
InChI Key KTZNVZJECQAMBV-UHFFFAOYSA-N
Synonym 1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine
PubChem CID 70768
IUPAC Name 1-(cyclohexen-1-yl)pyrrolidine
SMILES C1CCC(=CC1)N2CCCC2

Specifications

Density 0.94
Boiling Point 114°C to 115°C (15 mmHg)
Flash Point 66°C (151°F)
Refractive Index 1.5217
Quantity 5 g
Beilstein 114797
Sensitivity Air sensitive
Formula Weight 151.25
Percent Purity 97%
Chemical Name or Material 1-(1-Cyclohexen-1-yl)pyrrolidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.