missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diaminodecane, 97%, Thermo Scientific Chemicals

Product Code. 10540371
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-25-3
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.31
MDL Number MFCD00008151
InChI Key YQLZOAVZWJBZSY-UHFFFAOYSA-N
Synonym 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
PubChem CID 1317
IUPAC Name decane-1,10-diamine
SMILES C(CCCCCN)CCCCN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 62.0°C to 63.0°C
Color White to Yellow
Boiling Point 140.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)10NH2
Quantity 25 g
Beilstein 04, 273
Formula Weight 172.31
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 1, 10-Diaminodecane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,10-Diaminodecane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'