missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals

Product Code. 15417947
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23978-55-4
Molecular Formula C12H28N2O4
Molecular Weight (g/mol) 264.37
MDL Number MFCD00005112
InChI Key NLMDJJTUQPXZFG-UHFFFAOYSA-P
Synonym kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane
PubChem CID 72805
IUPAC Name 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
SMILES C1COCCOCC[NH2+]CCOCCOCC[NH2+]1

Specifications

Quantity 5 g
Beilstein 609764
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 262.35
Percent Purity 96%
Chemical Name or Material 1,10-Diaza-18-crown-6

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'