missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dibromodecane, 97%, Thermo Scientific Chemicals

Product Code. 10386220
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4101-68-2
Molecular Formula C10H20Br2
Molecular Weight (g/mol) 300.08
MDL Number MFCD00000222
InChI Key GTQHJCOHNAFHRE-UHFFFAOYSA-N
Synonym decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
PubChem CID 221483
IUPAC Name 1,10-dibromodecane
SMILES BrCCCCCCCCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 25°C to 30°C
Color Beige to Brown
Density 1.3400g/mL
Boiling Point 160°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)10Br
Refractive Index 1.4912
Quantity 25 mL
Beilstein 01, I, 64
Specific Gravity 1.34
Solubility Information Solubility in water: insoluble
Formula Weight 300.07
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1, 10-Dibromodecane
Show More Show Less
Product Identifier
  • 1,10-Dibromodecane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'