missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline-5,6-dione, 98%

Product Code. 15535186 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15535186

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27318-90-7 | C12H6N2O2 | 210.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27318-90-7
Molecular Formula C12H6N2O2
Molecular Weight (g/mol) 210.19
MDL Number MFCD00014473
InChI Key KCALAFIVPCAXJI-UHFFFAOYSA-N
Synonym 1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17
PubChem CID 72810
IUPAC Name 1,10-phenanthroline-5,6-dione
SMILES O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 257.0°C to 258.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material 1,10-Phenanthroline-5,6-dione
compliance-icons
Product Identifier
  • 1,10-Phenanthroline-5,6-dione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.