missing translation for 'onlineSavingsMsg'
Learn More
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals Product Code.: 15432088

1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals

Product Code. 15432088
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15432088

Brand: Thermo Scientific Alfa Aesar H55747.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

Solubility
Insoluble in water.

Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27318-90-7
Molecular Formula C12H6N2O2
Molecular Weight (g/mol) 210.19
MDL Number MFCD00014473
InChI Key KCALAFIVPCAXJI-UHFFFAOYSA-N
Synonym 1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17
PubChem CID 72810
IUPAC Name 1,10-phenanthroline-5,6-dione
SMILES O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 260°C (decomposition)
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material 1,10-Phenanthroline-5,6-dione

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ14J026 Certificate TypeCertificate of Analysis Date25/04/2023 Catalog NumberH55747.MD
Lot NumberQ14J027 Certificate TypeCertificate of Analysis Date25/04/2023 Catalog NumberH55747.MD
Lot NumberS27I036 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberH55747.03,H55747.06,H55747.MD
Lot Number5002K27R Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55747.03,H55747.06,H55747.MD
Lot NumberP09H003 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55747.03,H55747.06,H55747.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.