missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 99+%, Thermo Scientific Chemicals

64.12 € - 1620.06 €

Chemical Identifiers

CAS 66-71-7
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00011678
InChI Key DGEZNRSVGBDHLK-UHFFFAOYSA-N
Synonym o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline
PubChem CID 1318
ChEBI CHEBI:44975
IUPAC Name 1,10-phenanthroline
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10695972
View Documents
Thermo Scientific Acros
157530050
5 g
64.12 €
5g
Please to purchase this item. Need a web account? Register with us today!
10337620
View Documents
Thermo Scientific Acros
157530250
25 g
187.94 €
25g
Please to purchase this item. Need a web account? Register with us today!
10337481
View Documents
Thermo Scientific Acros
157531000
100 g
507.53 €
100g
Please to purchase this item. Need a web account? Register with us today!
15294644
View Documents
Thermo Scientific Acros
157535000
500 g
1620.06 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

66-71-7
180.21
DGEZNRSVGBDHLK-UHFFFAOYSA-N
1318
1,10-phenanthroline
C12H8N2
MFCD00011678
o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline
CHEBI:44975
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1

Specifications

66-71-7
100.0
Pink or White to Yellow
99+%
C12H8N2
23,227
o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline
DGEZNRSVGBDHLK-UHFFFAOYSA-N
1,10-phenanthroline
1318
180.21
Powder
99.0
117.0°C to 120.0°C
Authentic
Glass bottle
MFCD00011678
15,7328
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone, ether and benzene
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1
180.21
CHEBI:44975
99+%
1, 10-Phenanthroline
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-629-2

missing translation for 'rtecsNumber' : SF8300000

missing translation for 'tsca' : TSCA

Product Identifier
  • 1,10-Phenanthroline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only