missing translation for 'onlineSavingsMsg'
Learn More
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals Product Code.: 11387308

1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals

Product Code. 11387308
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11387308

Brand: Thermo Scientific Alfa Aesar 030562.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,10-Phenanthroline iron(II) sulfate is used as an indicator for the iron phenanthroline compound in analytical chemistry. Further, it is used as a redox indicator for making oscillatory Belousov-Zhabotinsky reactions visible.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
Synonym ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
PubChem CID 84567
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Color Red
Quantity 250 g
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
Formula Weight 692.53
Concentration or Composition (by Analyte or Components) 0.025M aq. soln.
Physical Form Liquid
Chemical Name or Material 1,10-Phenanthroline iron(II) sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM13L050 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number030562.30,030562.18
Lot NumberW13K040 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog Number030562.30,030562.18
Lot NumberT29K053 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number030562.30,030562.18
Lot NumberX04J014 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number030562.30,030562.18
Lot NumberN20J030 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number030562.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.