missing translation for 'onlineSavingsMsg'
Learn More
1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals Product Code.: 11488106

1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals

Product Code. 11488106
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11488106

Brand: Thermo Scientific Alfa Aesar A14140.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

Solubility
Soluble in ethanol, acetone and methanol. Slightly soluble in water and benzene.

Notes
Working solutions of 1,10-Phenanthroline monohydrate may stable for a few days at refrigerated temperatures. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
PubChem CID 21226
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 97°C to 101°C
Odor Odorless
Quantity 10 g
UN Number UN2811
Beilstein 4008096
Merck Index 14,7214
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
Solubility Information Soluble in ethanol,acetone and methanol. Slightly soluble in water and benzene.
IUPAC Name 1,10-phenanthroline;hydrate
Formula Weight 198.23 (180.21 Anhydrous)
Percent Purity ≥99%
Chemical Name or Material 1,10-Phenanthroline monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249860 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA14140.09,A14140.18,A14140.30
Lot Number10247254 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog NumberA14140.09,A14140.18,A14140,A14140.30
Lot NumberD06Y020 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14140.09,A14140.18,A14140.30
Lot Number10112653 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14140.09,A14140.18,A14140.30
Lot Number10127179 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14140.09,A14140.18,A14140.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.