Learn More
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals
Description
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 97°C to 101°C |
Quantity | 10 g |
UN Number | UN2811 |
Beilstein | 4008096 |
Merck Index | 14,7214 |
Solubility Information | Soluble in alcohol,acetone; Slightly soluble in waterSoluble in ethanol,acetone and methanol; Slightly soluble in water and benzene. |
Formula Weight | 198.23 (180.21 Anhydrous) |
Grade | ACS |
Physical Form | Crystalline |
Chemical Name or Material | 1, 10-Phenanthroline |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y14J046 | Certificate of Analysis | 05/12/2023 | 030910.18,030910.09 |
R23J023 | Certificate of Analysis | 06/06/2023 | 030910.09 |
W22I032 | Certificate of Analysis | 13/10/2022 | 030910.18,030910.09 |
S01I016 | Certificate of Analysis | 09/06/2022 | 030910.18,030910.09 |
M04I028 | Certificate of Analysis | 12/04/2022 | 030910.18,030910.09 |
Safety and Handling


- 1,10-Phenanthroline monohydrate
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- MIXTURE LIST-Contains : 1,10-Phenanthrolinemonohydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.