missing translation for 'onlineSavingsMsg'
Learn More
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals Product Code.: 15441177

1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals

Product Code. 15441177
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 15441177

Brand: Thermo Scientific Alfa Aesar 030910.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrateShow MoreShow Less
PubChem CID 21226
IUPAC Name 1,10-phenanthroline;hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 97°C to 101°C
Odor Odorless
Quantity 50 g
UN Number UN2811
Beilstein 4008096
Merck Index 14,7214
Solubility Information Soluble in alcohol,acetone; Slightly soluble in waterSoluble in ethanol,acetone and methanol; Slightly soluble in water and benzene.
Formula Weight 198.23 (180.21 Anhydrous)
Grade ACS
Physical Form Crystalline
Chemical Name or Material 1,10-Phenanthroline monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY14J046 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number030910.18,030910.09
Lot NumberW22I032 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number030910.18,030910.09
Lot NumberS01I016 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog Number030910.18,030910.09
Lot NumberM04I028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number030910.18,030910.09
Lot NumberJ24Y031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number030910.18,030910.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.