missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Monohydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10171910
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
100 g
Packaging:
Amber glass bottle
Plastic powder jar
Vial

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PubChem CID 21226
IUPAC Name 1,10-phenanthroline;hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 97°C
Color White
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 198.23g/mol
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • 1,10-Phenanthroline monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Phenanthroline (1, 10), monohydrate, 1,10-Phenanthroline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'